Product Details

SNP ID
rs148503247
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:2797817 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACCTGCAAACCAGAATATGCCTA[C/T]GGTTCAGCAGGCAGTCCTCCAAAGA
Phenotype
MIM: 600611
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FKBP4 PubMed Links

Gene Details

Gene
FKBP4
Gene Name
FK506 binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002014.3 537 Silent Mutation TAC,TAT Y113Y NP_002005.1
XM_011520929.2 537 Silent Mutation TAC,TAT Y68Y XP_011519231.1

View Full Product Details