Product Details

SNP ID
rs148770476
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:223145 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGTCACTCACCGTAAACCCAAGC[C/T]ACACAGAGGGACTCGAAGATGGCCA
Phenotype
MIM: 603080 MIM: 615097
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC6A12 PubMed Links

Gene Details

Gene
SLC6A12
Gene Name
solute carrier family 6 member 12
There are no transcripts associated with this gene.

Gene
SLC6A13
Gene Name
solute carrier family 6 member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190997.2 1559 Silent Mutation GTA,GTG V375V NP_001177926.1
NM_001243392.1 1559 Intron NP_001230321.1
NM_016615.4 1559 Silent Mutation GTA,GTG V467V NP_057699.2
XM_006719008.3 1559 Silent Mutation GTA,GTG V224V XP_006719071.1
XM_011521012.2 1559 Silent Mutation GTA,GTG V348V XP_011519314.1
XM_011521013.1 1559 Silent Mutation GTA,GTG V305V XP_011519315.1
XM_011521014.1 1559 Silent Mutation GTA,GTG V305V XP_011519316.1
XM_017019842.1 1559 Silent Mutation GTA,GTG V310V XP_016875331.1
XM_017019843.1 1559 Silent Mutation GTA,GTG V307V XP_016875332.1
XM_017019844.1 1559 Intron XP_016875333.1
XM_017019845.1 1559 Silent Mutation GTA,GTG V224V XP_016875334.1
XM_017019846.1 1559 Intron XP_016875335.1
XM_017019847.1 1559 Intron XP_016875336.1

View Full Product Details