Product Details

SNP ID
rs148775301
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56468614 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGATGGCGCTGCTTTTTGCACGTT[A/C]TTTGCGCTTGTGCCGCTGGGGAGCC
Phenotype
MIM: 606365
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GLS2 PubMed Links

Gene Details

Gene
GLS2
Gene Name
glutaminase 2
There are no transcripts associated with this gene.

Gene
SPRYD4
Gene Name
SPRY domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207344.3 98 Missense Mutation TAT,TCT Y8S NP_997227.1

View Full Product Details