Product Details

SNP ID
rs148955691
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10809211 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACATCTTCAGAAAAGCTTCCCTCA[A/G]CTTGCTATTTCCCATAATTAGAATG
Phenotype
MIM: 604793 MIM: 604794 MIM: 604795
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TAS2R7 PubMed Links

Gene Details

Gene
TAS2R7
Gene Name
taste 2 receptor member 7
There are no transcripts associated with this gene.

Gene
TAS2R8
Gene Name
taste 2 receptor member 8
There are no transcripts associated with this gene.

Gene
TAS2R9
Gene Name
taste 2 receptor member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023917.2 958 Silent Mutation CTG,TTG L289L NP_076406.1

View Full Product Details