Product Details

SNP ID
rs149091495
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:9833222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAATTATTTCTTGTTCCAATCATC[A/G]GAATCATTCCATTGAATGTGTCCCT
Phenotype
MIM: 605029
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLRF1 PubMed Links

Gene Details

Gene
KLRF1
Gene Name
killer cell lectin like receptor F1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291822.1 Intron NP_001278751.1
NM_001291823.1 Intron NP_001278752.1
NM_016523.2 Intron NP_057607.1
XM_017019414.1 Intron XP_016874903.1
XM_017019415.1 Intron XP_016874904.1

View Full Product Details