Product Details

SNP ID
rs149098520
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:51098806 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGATGAGCACATGAATTCCTGT[C/T]GGCCCCTGCTTGTAAATCTGGCTGA
Phenotype
MIM: 189889
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
TFCP2 PubMed Links

Gene Details

Gene
TFCP2
Gene Name
transcription factor CP2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173452.1 2110 Silent Mutation CCA,CCG P463P NP_001166923.1
NM_001173453.1 2110 Silent Mutation CCA,CCG P412P NP_001166924.1
NM_005653.4 2110 Silent Mutation CCA,CCG P463P NP_005644.2

View Full Product Details