Product Details

SNP ID
rs149122594
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:75042320 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGCCTTCCCCAAGTCATTAAGTA[A/T]GAGATCTGGCCTCGGCTTGCGTGTA
Phenotype
MIM: 176256
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KCNC2 PubMed Links

Gene Details

Gene
KCNC2
Gene Name
potassium voltage-gated channel subfamily C member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001260497.1 2231 UTR 3 NP_001247426.1
NM_001260498.1 2231 Intron NP_001247427.1
NM_001260499.1 2231 UTR 3 NP_001247428.1
NM_139136.3 2231 Missense Mutation CAT,CTT H612L NP_631874.1
NM_139137.3 2231 UTR 3 NP_631875.1
NM_153748.2 2231 Missense Mutation CAT,CTT H557L NP_715624.1
XM_005268855.3 2231 Intron XP_005268912.1
XM_006719382.3 2231 Missense Mutation CAT,CTT H612L XP_006719445.1
XM_006719383.3 2231 UTR 3 XP_006719446.1
XM_006719384.3 2231 UTR 3 XP_006719447.1
XM_006719385.3 2231 UTR 3 XP_006719448.1
XM_006719390.3 2231 Intron XP_006719453.1
XM_017019282.1 2231 Intron XP_016874771.1

View Full Product Details