Product Details

SNP ID
rs149226113
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:12622107 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTACTTTCTTCAGATACGGATTGG[A/G]TGTGTTAATAAGGACCTTATTGAGT
Phenotype
MIM: 603476
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CREBL2 PubMed Links

Gene Details

Gene
CREBL2
Gene Name
cAMP responsive element binding protein like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001310.3 Intron NP_001301.1

View Full Product Details