Product Details
- SNP ID
-
rs149248943
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:102076806 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAAACGGTACATTACCTGAATAAG[C/G]AGGCTCGATCCATGTGAACAGGTCT
- Phenotype
-
MIM: 609264
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NUP37
PubMed Links
Gene Details
- Gene
- NUP37
- Gene Name
- nucleoporin 37
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_024057.3 |
911 |
Missense Mutation |
TCC,TGC |
S255C |
NP_076962.2 |
View Full Product Details