Product Details
- SNP ID
-
rs149511139
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:56469559 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCTGCTGACCCATTCAGGGCTTGA[A/G]GTGCCCGAGGGCCTCTAGTATGTCC
- Phenotype
-
MIM: 606365
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GLS2
PubMed Links
Gene Details
- Gene
- GLS2
- Gene Name
- glutaminase 2
There are no transcripts associated with this gene.
- Gene
- SPRYD4
- Gene Name
- SPRY domain containing 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_207344.3 |
681 |
Silent Mutation |
GAA,GAG |
E202E |
NP_997227.1 |
View Full Product Details