Product Details

SNP ID
rs149555390
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53252972 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTTCGGTTTCAACTGGACTTCT[A/G]TCAGGTCTACTTCCTGGCCCTGGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MFSD5 PubMed Links

Gene Details

Gene
MFSD5
Gene Name
major facilitator superfamily domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170790.1 282 Missense Mutation TAT,TGT Y153C NP_001164261.1
NM_032889.4 282 Missense Mutation TAT,TGT Y46C NP_116278.3
XM_005269197.1 282 Missense Mutation TAT,TGT Y153C XP_005269254.1
XM_005269198.4 282 Missense Mutation TAT,TGT Y46C XP_005269255.1

View Full Product Details