Product Details

SNP ID
rs149601103
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:89348883 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCATTGAAGACAATTATATAGT[C/T]AGTTCCAGATGCAAAAGGAAACAGC
Phenotype
MIM: 602748
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DUSP6 PubMed Links

Gene Details

Gene
DUSP6
Gene Name
dual specificity phosphatase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001946.3 1899 UTR 3 NP_001937.2
NM_022652.3 1899 UTR 3 NP_073143.2
Gene
LOC105369889
Gene Name
uncharacterized LOC105369889
There are no transcripts associated with this gene.

View Full Product Details