Product Details

SNP ID
rs149651954
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:122781327 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCTTCTCTTCTCTCTGAAGACCT[C/T]GAGGTTGGGATTAGTTTTTGATAAG
Phenotype
MIM: 613481
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC62 PubMed Links

Gene Details

Gene
CCDC62
Gene Name
coiled-coil domain containing 62
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_201435.4 1443 Silent Mutation CTC,CTT L131L NP_958843.2
XM_006719643.1 1443 Silent Mutation CTC,CTT L131L XP_006719706.1
XM_006719644.1 1443 Silent Mutation CTC,CTT L131L XP_006719707.1
XM_011538856.1 1443 Silent Mutation CTC,CTT L131L XP_011537158.1
XM_011538857.1 1443 Silent Mutation CTC,CTT L131L XP_011537159.1
XM_011538859.1 1443 Intron XP_011537161.1
XM_017020033.1 1443 Silent Mutation CTC,CTT L131L XP_016875522.1

View Full Product Details