Product Details

SNP ID
rs149722587
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:14506214 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGTTAGGTGAGTTCAGGTCCTCA[C/G]GGCAGCAGATGGTATTACAGGGGTC
Phenotype
MIM: 613644
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATF7IP PubMed Links

Gene Details

Gene
ATF7IP
Gene Name
activating transcription factor 7 interacting protein
There are no transcripts associated with this gene.

Gene
PLBD1
Gene Name
phospholipase B domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024829.5 1588 Missense Mutation CCT,CGT P476R NP_079105.4

View Full Product Details