Product Details

SNP ID
rs149731216
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:104457461 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGGATGAACAGAATCTGCCGGA[C/T]GGTGCTGGCCACTTGCTTGGGATCC
Phenotype
MIM: 610128
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHST11 PubMed Links

Gene Details

Gene
CHST11
Gene Name
carbohydrate (chondroitin 4) sulfotransferase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173982.1 548 Missense Mutation ACG,ATG T17M NP_001167453.1
NM_018413.5 548 Missense Mutation ACG,ATG T17M NP_060883.1
XM_017019369.1 548 Intron XP_016874858.1

View Full Product Details