Product Details

SNP ID
rs149731918
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:112406805 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGGTCCCGGGGGTAATGCTGGCT[C/T]GCAGTTTTCTCACGTGCTGACTGAA
Phenotype
MIM: 603703
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPL6 PubMed Links

Gene Details

Gene
RPL6
Gene Name
ribosomal protein L6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000970.4 701 Intron NP_000961.2
NM_001024662.2 701 Intron NP_001019833.1
NM_001320137.1 701 Intron NP_001307066.1
NM_001320138.1 701 Missense Mutation CAA,CGA Q141R NP_001307067.1
NM_001320139.1 701 Intron NP_001307068.1
NM_001320140.1 701 Missense Mutation CAA,CGA Q141R NP_001307069.1
NM_001320141.1 701 Missense Mutation CAA,CGA Q141R NP_001307070.1
NM_001320142.1 701 Intron NP_001307071.1
XM_017019781.1 701 Missense Mutation CAA,CGA Q141R XP_016875270.1
XM_017019782.1 701 Intron XP_016875271.1

View Full Product Details