Product Details

SNP ID
rs149961727
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:122205959 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGAATGTCAGTTCCACACTTAG[C/T]GAATTTGATCCAAGCTAAACTGAGT
Phenotype
MIM: 605864 MIM: 605219
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
B3GNT4 PubMed Links

Gene Details

Gene
B3GNT4
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011538732.1 Intron XP_011537034.1
Gene
DIABLO
Gene Name
diablo IAP-binding mitochondrial protein
There are no transcripts associated with this gene.

Gene
LRRC43
Gene Name
leucine rich repeat containing 43
There are no transcripts associated with this gene.

View Full Product Details