Product Details

SNP ID
rs149961969
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:92143637 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTCTTGTAAAATTTCCAGTACTA[A/C]CATGTTTAAAACGTTTAACTTTCCT
Phenotype
MIM: 109580
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BTG1 PubMed Links

Gene Details

Gene
BTG1
Gene Name
BTG anti-proliferation factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001731.2 1321 UTR 3 NP_001722.1
Gene
LINC01619
Gene Name
long intergenic non-protein coding RNA 1619
There are no transcripts associated with this gene.

Gene
LOC101928617
Gene Name
uncharacterized LOC101928617
There are no transcripts associated with this gene.

View Full Product Details