Product Details

SNP ID
rs150056191
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53940922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGGCCTGAAATTCCTCTGCATAC[A/G]GAATTGGAAGAAGAGGTAGGTGGTT
Phenotype
MIM: 142976
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HOXC13 PubMed Links

Gene Details

Gene
HOXC13
Gene Name
homeobox C13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017410.2 Intron NP_059106.2
Gene
HOXC13-AS
Gene Name
HOXC13 antisense RNA
There are no transcripts associated with this gene.

View Full Product Details