Product Details

SNP ID
rs150140900
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:7304476 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATAAATCGCTGTAACAGGCCATTG[C/T]CTAAAAACTTTAACTTTGCTGCAGA
Phenotype
MIM: 614360
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACSM4 PubMed Links

Gene Details

Gene
ACSM4
Gene Name
acyl-CoA synthetase medium-chain family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080454.1 145 Missense Mutation CCT,TCT P49S NP_001073923.1

View Full Product Details