Product Details
- SNP ID
-
rs150443540
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:2858923 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAGGAGCCTTTGCGGTGATTCAAG[C/G]GGGGGAGCACTTTGCAAGGGAGTGG
- Phenotype
-
MIM: 602341
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FOXM1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs28919869] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FOXM1
- Gene Name
- forkhead box M1
- Gene
- LOC100507424
- Gene Name
- uncharacterized LOC100507424
There are no transcripts associated with this gene.
- Gene
- LOC101929469
- Gene Name
- uncharacterized protein ENSP00000372125
There are no transcripts associated with this gene.
View Full Product Details