Product Details

SNP ID
rs150474647
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53985395 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACTTCAATTGCGGGGTGATGAGG[A/G]GCTGCGGGCTCGCGCCCTCGCTCTC
Phenotype
MIM: 605560 MIM: 605559 MIM: 609687
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HOXC-AS2 PubMed Links

Gene Details

Gene
HOXC-AS2
Gene Name
HOXC cluster antisense RNA 2
There are no transcripts associated with this gene.

Gene
HOXC-AS3
Gene Name
HOXC cluster antisense RNA 3
There are no transcripts associated with this gene.

Gene
HOXC10
Gene Name
homeobox C10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017409.3 234 Missense Mutation AGC,GGC S46G NP_059105.2
Gene
HOXC11
Gene Name
homeobox C11
There are no transcripts associated with this gene.

Gene
MIR196A2
Gene Name
microRNA 196a-2
There are no transcripts associated with this gene.

View Full Product Details