Product Details

SNP ID
rs150655870
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:69350257 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCCCAGGAGCAGTTAATGCCTGT[A/C]ATTTATCCTGCAGTGGTAAGACAAG
Phenotype
MIM: 153450 MIM: 602116
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LYZ PubMed Links

Gene Details

Gene
LYZ
Gene Name
lysozyme
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000239.2 341 Missense Mutation AAT,CAT N96H NP_000230.1
Gene
YEATS4
Gene Name
YEATS domain containing 4
There are no transcripts associated with this gene.

View Full Product Details