Product Details

SNP ID
rs150772647
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53504402 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGTGCCTTTTCCTTCAGGAGCC[C/G]CCCCATGGAACTGCAGCCCCCTGTC
Phenotype
MIM: 606371 MIM: 600447 MIM: 604643 MIM: 605053
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATF7 PubMed Links

Gene Details

Gene
ATF7
Gene Name
activating transcription factor 7
There are no transcripts associated with this gene.

Gene
LOC100652999
Gene Name
uncharacterized LOC100652999
There are no transcripts associated with this gene.

Gene
MAP3K12
Gene Name
mitogen-activated protein kinase kinase kinase 12
There are no transcripts associated with this gene.

Gene
NPFF
Gene Name
neuropeptide FF-amide peptide precursor
There are no transcripts associated with this gene.

Gene
TARBP2
Gene Name
TARBP2, RISC loading complex RNA binding subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004178.4 535 Missense Mutation CCC,CGC P122R NP_004169.3
NM_134323.1 535 Missense Mutation CCC,CGC P143R NP_599150.1
NM_134324.2 535 Missense Mutation CCC,CGC P122R NP_599151.2
XM_005269114.1 535 Missense Mutation CCC,CGC P122R XP_005269171.1
XM_005269115.2 535 Missense Mutation CCC,CGC P122R XP_005269172.1
XM_005269117.1 535 Missense Mutation CCC,CGC P143R XP_005269174.1
XM_005269120.4 535 UTR 5 XP_005269177.1
XM_005269122.3 535 UTR 5 XP_005269179.1
XM_006719581.1 535 Missense Mutation CCC,CGC P44R XP_006719644.1
XM_011538712.2 535 UTR 5 XP_011537014.1
XM_017019910.1 535 Missense Mutation CCC,CGC P122R XP_016875399.1
XM_017019911.1 535 UTR 5 XP_016875400.1

View Full Product Details