Product Details

SNP ID
rs150945344
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:8995138 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTAGGCTCCAACTACTCCACTTG[C/T]GCCAGCTGTCCTAGCTGCCCAGACC
Phenotype
MIM: 604874
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLRG1 PubMed Links

Gene Details

Gene
KLRG1
Gene Name
killer cell lectin like receptor G1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005810.3 763 Silent Mutation TGC,TGT C69C NP_005801.3
XM_017018682.1 763 Silent Mutation TGC,TGT C69C XP_016874171.1
XM_017018683.1 763 Silent Mutation TGC,TGT C69C XP_016874172.1
XM_017018684.1 763 Silent Mutation TGC,TGT C69C XP_016874173.1
XM_017018685.1 763 Silent Mutation TGC,TGT C69C XP_016874174.1

View Full Product Details