Product Details

SNP ID
rs150987658
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:4810257 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACGCTGCTCGGAGACCCTGGCCG[A/G]CGAGTCCGCTTCTTCGACCCCCTGA
Phenotype
MIM: 176257
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNA6 PubMed Links

Gene Details

Gene
KCNA6
Gene Name
potassium voltage-gated channel subfamily A member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002235.3 1082 Silent Mutation CGA,CGG R72R NP_002226.1
XM_005253686.3 1082 Silent Mutation CGA,CGG R72R XP_005253743.1
XM_011520955.2 1082 Silent Mutation CGA,CGG R72R XP_011519257.1
XM_017019270.1 1082 Silent Mutation CGA,CGG R72R XP_016874759.1
XM_017019271.1 1082 Silent Mutation CGA,CGG R72R XP_016874760.1
XM_017019272.1 1082 Silent Mutation CGA,CGG R72R XP_016874761.1

View Full Product Details