Product Details

SNP ID
rs151134796
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:75480890 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGCCAGACAAAGCATGCGTGTC[A/G]CACTTGCTACAATAGCCTGGATGGT
Phenotype
MIM: 602692
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLIPR1 PubMed Links

Gene Details

Gene
GLIPR1
Gene Name
GLI pathogenesis related 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006851.2 137 Missense Mutation ACA,GCA T4A NP_006842.2
XM_011537775.2 137 Missense Mutation ACA,GCA T4A XP_011536077.1
XM_017018721.1 137 Intron XP_016874210.1

View Full Product Details