Product Details

SNP ID
rs151268143
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:98614086 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAATGATATTACATCAGTTACTAA[A/C]CCTGAAATCCGTCGTATATCTGTTT
Phenotype
MIM: 609861
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IKBIP PubMed Links

Gene Details

Gene
IKBIP
Gene Name
IKBKB interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153687.3 784 Intron NP_710154.1
NM_201612.2 784 Silent Mutation GGG,GGT G184G NP_963906.1
NM_201613.2 784 UTR 3 NP_963907.1

View Full Product Details