Product Details

SNP ID
rs151331299
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:106781139 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGTTTAACTCTTTTTTTTTAAAT[A/G]GGGTTTTATCGTGTTGCCCTGGCTG
Phenotype
MIM: 609147
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LOC100287944 PubMed Links

Gene Details

Gene
LOC100287944
Gene Name
uncharacterized LOC100287944
There are no transcripts associated with this gene.

Gene
RIC8B
Gene Name
RIC8 guanine nucleotide exchange factor B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005268998.3 Intron XP_005269055.1
XM_011538526.2 Intron XP_011536828.1
XM_011538527.2 Intron XP_011536829.1
XM_011538528.2 Intron XP_011536830.1
XM_011538529.2 Intron XP_011536831.1
XM_017019548.1 Intron XP_016875037.1
XM_017019549.1 Intron XP_016875038.1

View Full Product Details