Product Details

SNP ID
rs151334598
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:107732871 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGAGACTACCAGAGGAATCCTG[C/T]ATTTTAAAAAAGTTTAATTTTAATC
Phenotype
MIM: 605780
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101929162 PubMed Links

Gene Details

Gene
LOC101929162
Gene Name
uncharacterized LOC101929162
There are no transcripts associated with this gene.

Gene
PRDM4
Gene Name
PR domain 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012406.3 4182 UTR 3 NP_036538.3
XM_005268593.1 4182 UTR 3 XP_005268650.1

View Full Product Details