Product Details

SNP ID
rs138409614
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:113208634 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTGGTTAATGGTAATGTGCGCCA[C/T]GGGAGCGCCGCCGAACGGAGAGCGC
Phenotype
MIM: 603137 MIM: 613713
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CUL4A PubMed Links

Gene Details

Gene
CUL4A
Gene Name
cullin 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008895.2 15 Intron NP_001008895.1
NM_001278513.1 15 Intron NP_001265442.1
NM_001278514.1 15 Intron NP_001265443.1
NM_003589.2 15 Intron NP_003580.1
XM_011537523.2 15 Intron XP_011535825.1
Gene
PCID2
Gene Name
PCI domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127202.3 15 Missense Mutation NP_001120674.1
NM_001127203.3 15 Missense Mutation NP_001120675.1
NM_001258213.2 15 UTR 5 NP_001245142.1
NM_001320655.1 15 UTR 5 NP_001307584.1
NM_001320656.1 15 Missense Mutation NP_001307585.1
NM_001320657.1 15 Missense Mutation NP_001307586.1
NM_001320659.1 15 Missense Mutation NP_001307588.1
NM_001320660.1 15 UTR 5 NP_001307589.1
NM_018386.4 15 Missense Mutation NP_060856.2
XM_005268325.3 15 Missense Mutation XP_005268382.1
XM_017020664.1 15 Intron XP_016876153.1
XM_017020665.1 15 Intron XP_016876154.1
XM_017020666.1 15 UTR 5 XP_016876155.1
XM_017020667.1 15 UTR 5 XP_016876156.1
XM_017020668.1 15 Intron XP_016876157.1
XM_017020669.1 15 UTR 5 XP_016876158.1
XM_017020670.1 15 Intron XP_016876159.1
XM_017020671.1 15 Intron XP_016876160.1
XM_017020672.1 15 Intron XP_016876161.1

View Full Product Details