Product Details

SNP ID
rs138728859
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:94709894 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATATACATTTAAAAAGAAACAAAG[C/G]TAATAATTGAAGGTTCCTTTTTTCT
Phenotype
MIM: 604974
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC101927248 PubMed Links

Gene Details

Gene
LOC101927248
Gene Name
uncharacterized LOC101927248
There are no transcripts associated with this gene.

Gene
SOX21
Gene Name
SRY-box 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007084.3 2650 UTR 3 NP_009015.1
Gene
SOX21-AS1
Gene Name
SOX21 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details