Product Details

SNP ID
rs139004069
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:114242237 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTTGATGTCTACTGTTTTGAAGC[A/G]TTCGATCTTTTAACATCACATCACA
Phenotype
MIM: 603461
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDC16 PubMed Links

Gene Details

Gene
CDC16
Gene Name
cell division cycle 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005266206.1 683 Silent Mutation GCA,GCG A166A XP_005266263.1
XM_006719992.1 683 Silent Mutation GCA,GCG A165A XP_006720055.1
XM_006719993.1 683 Silent Mutation GCA,GCG A72A XP_006720056.1
XM_011534851.1 683 Silent Mutation GCA,GCG A166A XP_011533153.1
XM_011534853.1 683 Silent Mutation GCA,GCG A166A XP_011533155.1
XM_017020830.1 683 Silent Mutation GCA,GCG A166A XP_016876319.1
XM_017020831.1 683 Silent Mutation GCA,GCG A166A XP_016876320.1
XM_017020832.1 683 Silent Mutation GCA,GCG A72A XP_016876321.1
XM_017020833.1 683 Silent Mutation GCA,GCG A72A XP_016876322.1
Gene
MIR548AR
Gene Name
microRNA 548ar
There are no transcripts associated with this gene.

View Full Product Details