Product Details

SNP ID
rs139754496
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:46834930 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTAGAGCAACCATTGAGCAGTC[A/T]TTATCTGTTGTCTTGGCATCTTGCT
Phenotype
MIM: 182135
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HTR2A PubMed Links

Gene Details

Gene
HTR2A
Gene Name
5-hydroxytryptamine receptor 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000621.4 1314 Silent Mutation AAA,AAT K441N NP_000612.1
NM_001165947.2 1314 Silent Mutation AAA,AAT K357N NP_001159419.1

View Full Product Details