Product Details

SNP ID
rs139802434
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:19642529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATATTTTCCTTTAAGTTAAGAGT[A/G]AGGTCAAGGTGGAGAAAAAAGAGAC
Phenotype
MIM: 611626
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MPHOSPH8 PubMed Links

Gene Details

Gene
MPHOSPH8
Gene Name
M-phase phosphoprotein 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017520.3 Intron NP_059990.2
XM_011535124.2 Intron XP_011533426.1

View Full Product Details