Product Details

SNP ID
rs139849768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:24884049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTTTTAACAGTCTGGTCAGGAAA[C/T]GTGATTTCTTTTCTTCCATCTGGGT
Phenotype
MIM: 609279 MIM: 605793
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CENPJ PubMed Links

Gene Details

Gene
CENPJ
Gene Name
centromere protein J
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018451.4 3933 Silent Mutation ACA,ACG T1246T NP_060921.3
XM_011535149.2 3933 Intron XP_011533451.1
XM_011535150.2 3933 Intron XP_011533452.1
XM_017020673.1 3933 Intron XP_016876162.1
Gene
RNF17
Gene Name
ring finger protein 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184993.1 3933 Intron NP_001171922.1
NM_031277.2 3933 Intron NP_112567.2
XM_006719846.3 3933 Intron XP_006719909.1
XM_006719849.2 3933 Intron XP_006719912.1
XM_006719852.2 3933 Intron XP_006719915.1
XM_011535152.2 3933 Intron XP_011533454.1
XM_011535155.2 3933 Intron XP_011533457.1
XM_011535156.2 3933 Intron XP_011533458.1
XM_011535157.2 3933 Intron XP_011533459.1
XM_011535158.2 3933 Intron XP_011533460.1
XM_011535159.2 3933 Intron XP_011533461.1
XM_011535160.2 3933 Intron XP_011533462.1
XM_011535162.1 3933 Intron XP_011533464.1
XM_011535163.1 3933 Intron XP_011533465.1
XM_011535164.2 3933 Intron XP_011533466.1
XM_017020676.1 3933 Intron XP_016876165.1

View Full Product Details