Product Details

SNP ID
rs140397494
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:28662489 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGCAAGTGGACCTTTTGAAAGTC[A/C]TGATCTTCTTCGGAAAGGGTATATG
Phenotype
MIM: 613386
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
POMP PubMed Links

Gene Details

Gene
POMP
Gene Name
proteasome maturation protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015932.5 264 Missense Mutation CAT,CCT H28P NP_057016.1

View Full Product Details