Product Details

SNP ID
rs140432362
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:75525403 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCATGCACAGTTACAGAAGGCA[C/G]GTTTGCTGTTTTAACCCAGATAGAG
Phenotype
MIM: 612377
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
COMMD6 PubMed Links

Gene Details

Gene
COMMD6
Gene Name
COMM domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287392.1 1550 UTR 3 NP_001274321.1
NM_001287393.1 1550 UTR 3 NP_001274322.1
NM_001287394.1 1550 UTR 3 NP_001274323.1
NM_203495.3 1550 UTR 3 NP_987091.1
NM_203497.3 1550 UTR 3 NP_987093.1
XM_011534948.2 1550 Intron XP_011533250.1

View Full Product Details