Product Details
- SNP ID
-
rs140716826
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:43214119 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAGCATTTCTATTTCATTTGCAGA[C/T]ATCTGTTAGAAAGGAAGGAAAGTCA
- Phenotype
-
MIM: 610914
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ENOX1
PubMed Links
Gene Details
- Gene
- ENOX1
- Gene Name
- ecto-NOX disulfide-thiol exchanger 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001127615.1 |
2416 |
Missense Mutation |
ATA,ATG |
I601M |
NP_001121087.1 |
NM_001242863.1 |
2416 |
Missense Mutation |
ATA,ATG |
I601M |
NP_001229792.1 |
NM_017993.3 |
2416 |
Missense Mutation |
ATA,ATG |
I601M |
NP_060463.2 |
XM_005266439.3 |
2416 |
Missense Mutation |
ATA,ATG |
I601M |
XP_005266496.1 |
XM_011535126.2 |
2416 |
Missense Mutation |
ATA,ATG |
I601M |
XP_011533428.1 |
XM_011535127.2 |
2416 |
Missense Mutation |
ATA,ATG |
I601M |
XP_011533429.1 |
XM_011535128.2 |
2416 |
Missense Mutation |
ATA,ATG |
I601M |
XP_011533430.1 |
XM_011535132.2 |
2416 |
Missense Mutation |
ATA,ATG |
I562M |
XP_011533434.1 |
XM_017020637.1 |
2416 |
Missense Mutation |
ATA,ATG |
I601M |
XP_016876126.1 |
XM_017020638.1 |
2416 |
Missense Mutation |
ATA,ATG |
I601M |
XP_016876127.1 |
XM_017020639.1 |
2416 |
Missense Mutation |
ATA,ATG |
I601M |
XP_016876128.1 |
XM_017020640.1 |
2416 |
Missense Mutation |
ATA,ATG |
I562M |
XP_016876129.1 |
XM_017020641.1 |
2416 |
Intron |
|
|
XP_016876130.1 |
XM_017020642.1 |
2416 |
Intron |
|
|
XP_016876131.1 |
View Full Product Details