Product Details

SNP ID
rs141831246
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:38687584 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTGCTGGCGAACCGCGGACTCCG[A/G]GTGCCTTTCGGCCGTGAAGTCTGGC
Phenotype
MIM: 608945
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FREM2 PubMed Links

Gene Details

Gene
FREM2
Gene Name
FRAS1 related extracellular matrix protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207361.5 549 Silent Mutation CGA,CGG R80R NP_997244.4
XM_017020554.1 549 Silent Mutation CGA,CGG R80R XP_016876043.1
Gene
LINC00437
Gene Name
long intergenic non-protein coding RNA 437
There are no transcripts associated with this gene.

View Full Product Details