Product Details

SNP ID
rs141929561
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:20188291 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGACATACCATTTACATATTACA[A/G]TCTGAGCCTCTGAAACAGGGGGAAC
Phenotype
MIM: 121011
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GJB2 PubMed Links

Gene Details

Gene
GJB2
Gene Name
gap junction protein beta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004004.5 1497 UTR 3 NP_003995.2
XM_011535049.2 1497 UTR 3 XP_011533351.1

View Full Product Details