Product Details

SNP ID
rs142245737
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:110641641 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGAGTGAGGTCCAACTCTGAGC[A/G]GACACCACGTCATGTGGCACAGGGG
Phenotype
MIM: 612800 MIM: 615910
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CARS2 PubMed Links

Gene Details

Gene
CARS2
Gene Name
cysteinyl-tRNA synthetase 2, mitochondrial (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024537.3 Intron NP_078813.1
XM_006719953.2 Intron XP_006720016.1
XM_011521116.1 Intron XP_011519418.1
XM_011521117.2 Intron XP_011519419.1
XM_011521118.2 Intron XP_011519420.1
XM_017020741.1 Intron XP_016876230.1
XM_017020742.1 Intron XP_016876231.1
Gene
NAXD
Gene Name
NAD(P)HX dehydratase
There are no transcripts associated with this gene.

View Full Product Details