Product Details

SNP ID
rs142656048
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:49915102 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCAAATAATATTTTTTCAAAACC[A/C]GGTGGAGGCGTATGATAAAACTCAC
Phenotype
MIM: 607866
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SPRYD7 PubMed Links

Gene Details

Gene
SPRYD7
Gene Name
SPRY domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127482.2 818 Silent Mutation CCG,CCT P145P NP_001120954.1
NM_020456.3 818 Silent Mutation CCG,CCT P184P NP_065189.1
XM_017020677.1 818 Intron XP_016876166.1

View Full Product Details