Product Details

SNP ID
rs142834675
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:112489688 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCTTCCTCTGCTGCCGTCACTC[C/T]CCACTGGCCCTGAACAATCAAAAGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TUBGCP3 PubMed Links

Gene Details

Gene
TUBGCP3
Gene Name
tubulin gamma complex associated protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286277.1 3396 Missense Mutation AGA,GGA R810G NP_001273206.1
NM_001286278.1 3396 Intron NP_001273207.1
NM_001286279.1 3396 Intron NP_001273208.1
NM_006322.5 3396 Missense Mutation AGA,GGA R820G NP_006313.1
XM_005268293.1 3396 Intron XP_005268350.1
XM_011537462.2 3396 Missense Mutation AGA,GGA R720G XP_011535764.1
XM_011537463.2 3396 Missense Mutation AGA,GGA R591G XP_011535765.1
XM_011537464.2 3396 Missense Mutation AGA,GGA R591G XP_011535766.1
XM_017020323.1 3396 Missense Mutation AGA,GGA R591G XP_016875812.1

View Full Product Details