Product Details

SNP ID
rs144035947
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:99295447 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTTTGTTTACACCAGATTTCTC[A/G]GTGAGTGGGTTTTGTTTGGCAGTTC
Phenotype
MIM: 605741
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR183 PubMed Links

Gene Details

Gene
GPR183
Gene Name
G protein-coupled receptor 183
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004951.4 873 Silent Mutation ACC,ACT T233T NP_004942.1
XM_017020405.1 873 Silent Mutation ACC,ACT T233T XP_016875894.1
Gene
UBAC2
Gene Name
UBA domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144072.1 873 Intron NP_001137544.1
NM_177967.3 873 Intron NP_808882.1
XM_006719948.3 873 Intron XP_006720011.1
XM_011521082.2 873 Intron XP_011519384.1
XM_011521083.2 873 Intron XP_011519385.1
XM_011521084.2 873 Intron XP_011519386.1
XM_017020553.1 873 Intron XP_016876042.1

View Full Product Details