Product Details

SNP ID
rs144762909
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:77051817 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAAAATGTTCTGCCTACCTGAGC[C/T]CTGGAAACATCAGAACAGGCACCAC
Phenotype
MIM: 610392
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYCBP2 PubMed Links

Gene Details

Gene
MYCBP2
Gene Name
MYC binding protein 2, E3 ubiquitin protein ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015057.4 13884 Silent Mutation NP_055872.4
XM_005266299.2 13884 Missense Mutation XP_005266356.1
XM_006719779.2 13884 Missense Mutation XP_006719842.1
XM_006719780.2 13884 Missense Mutation XP_006719843.1
XM_006719781.2 13884 Missense Mutation XP_006719844.1
XM_006719782.2 13884 Missense Mutation XP_006719845.1
XM_006719783.2 13884 Missense Mutation XP_006719846.1
XM_006719784.2 13884 Missense Mutation XP_006719847.1
XM_006719785.2 13884 Missense Mutation XP_006719848.1
XM_006719786.2 13884 Silent Mutation XP_006719849.1
XM_006719787.2 13884 Missense Mutation XP_006719850.1
XM_006719788.2 13884 Missense Mutation XP_006719851.1
XM_006719789.2 13884 Missense Mutation XP_006719852.1
XM_011535004.2 13884 Missense Mutation XP_011533306.1
XM_011535005.2 13884 Missense Mutation XP_011533307.1
XM_017020454.1 13884 Missense Mutation XP_016875943.1
XM_017020455.1 13884 Missense Mutation XP_016875944.1
XM_017020456.1 13884 Missense Mutation XP_016875945.1
XM_017020457.1 13884 Missense Mutation XP_016875946.1
XM_017020458.1 13884 Missense Mutation XP_016875947.1
XM_017020459.1 13884 Missense Mutation XP_016875948.1
XM_017020460.1 13884 Missense Mutation XP_016875949.1
XM_017020461.1 13884 Missense Mutation XP_016875950.1
XM_017020462.1 13884 Missense Mutation XP_016875951.1
XM_017020463.1 13884 Missense Mutation XP_016875952.1
XM_017020464.1 13884 Missense Mutation XP_016875953.1
XM_017020465.1 13884 Missense Mutation XP_016875954.1
XM_017020466.1 13884 Missense Mutation XP_016875955.1
XM_017020467.1 13884 Missense Mutation XP_016875956.1
XM_017020468.1 13884 Missense Mutation XP_016875957.1

View Full Product Details