Product Details

SNP ID
rs145783562
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:46343457 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATCCTCGCACACCGGGGGCACTC[A/G]GAGGACTGGAAGCACTGTTTGTGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIAA0226L PubMed Links

Gene Details

Gene
KIAA0226L
Gene Name
KIAA0226 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286761.1 1865 Silent Mutation TCC,TCT S639S NP_001273690.1
NM_001286762.1 1865 Missense Mutation CCG,CTG P588L NP_001273691.1
NM_001286763.1 1865 Silent Mutation TCC,TCT S572S NP_001273692.1
NM_001286764.1 1865 Silent Mutation TCC,TCT S504S NP_001273693.1
NM_001286765.1 1865 Silent Mutation TCC,TCT S482S NP_001273694.1
NM_001286766.1 1865 Silent Mutation TCC,TCT S424S NP_001273695.1
NM_025113.3 1865 Silent Mutation TCC,TCT S639S NP_079389.2

View Full Product Details