Product Details

SNP ID
rs146576407
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:69706651 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAGAGCTGCACTGCACTCCTAGA[C/T]GGAAACTATTTATTTGAACAGTATC
Phenotype
MIM: 605332
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLHL1 PubMed Links

Gene Details

Gene
KLHL1
Gene Name
kelch like family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286725.1 Intron NP_001273654.1
NM_020866.2 Intron NP_065917.1
XM_017020678.1 Intron XP_016876167.1
XM_017020679.1 Intron XP_016876168.1

View Full Product Details