Product Details

SNP ID
rs146850453
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:49630937 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCTCACTGGGGAGGGGCTGCCC[A/G]AGGCCCTGCAGAGCCTGTGGAGCCT
Phenotype
MIM: 609351
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARL11 PubMed Links

Gene Details

Gene
ARL11
Gene Name
ADP ribosylation factor like GTPase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138450.5 734 Missense Mutation AAG,GAG K164E NP_612459.1
XM_005266253.3 734 Missense Mutation AAG,GAG K164E XP_005266310.1

View Full Product Details