Product Details

SNP ID
rs146856401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:42786827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGGATTTACAACTCCAGGCCCC[A/G]GTGGGAGGCCCTGCAGACCCGCTAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM216B PubMed Links

Gene Details

Gene
FAM216B
Gene Name
family with sequence similarity 216 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318932.1 287 Missense Mutation CAG,CGG Q55R NP_001305861.1
NM_182508.2 287 Missense Mutation CAG,CGG Q55R NP_872314.1

View Full Product Details